Learn More
CD4 Antibody (296712), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2410F
Product Code. 18618703
  • ONLINE EXCLUSIVE PRICE
    € 594.00
    € 561.75 / 0.10mL
    Save € 32.25
    5% Off
Description
CD4 Monoclonal antibody specifically detects CD4 in Canine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4
Monoclonal
FITC
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Canine
Purified
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
296712
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse myeloma cell line NS0-derived recombinant canine CD4, Val25-Lys401, Accession # P33705
0.1 mL
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
920
Store at 4°C in the dark.
IgG2b