missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (695542) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6676JF525
This item is not returnable.
View return policy
Description
CD4 Monoclonal antibody specifically detects CD4 in Cotton Rat samples. It is validated for Flow Cytometry,CyTOF
Specifications
| CD4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Mouse myeloma cell line NS0-derived recombinant cotton rat CD4, Lys26-Pro393 | |
| RUO | |
| Primary | |
| Cotton Rat | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 695542 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction