missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (C4/206), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
€ 552.00 - € 595.00
Specifications
| Antigen | CD4 |
|---|---|
| Clone | C4/206 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610316
|
Novus Biologicals
NBP2-47674B-100ul |
100 μg |
€ 595.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690416
|
Novus Biologicals
NBP2-47674B |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for ELISASpecifications
| CD4 | |
| ELISA | |
| Biotin | |
| Mouse | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 920 | |
| IgG1 κ | |
| Protein A or G purified |
| C4/206 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Recombinant full-length human CD4 protein (Uniprot: P01730) | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title