Learn More
CD4 Antibody (CE9.1 (Clenoliximab)), DyLight 350 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52681UV
Product Code. 18740213
  • € 620.00 / 0.10mL
Description
CD4 Monoclonal antibody specifically detects CD4 in Human, Chimpanzee samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Surface Plasmon Resonance, CyTOF, Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4
Monoclonal
DyLight 350
CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Domain 1 of human CD4.
0.1 mL
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
920
Store at 4°C in the dark.
IgG λ
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Surface Plasmon Resonance
CE9.1 (Clenoliximab)
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human, Chimpanzee