Learn More
CD4 Antibody (CL0395) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52928C
Product Code. 30298609
  • € 748.00 / 0.10mL
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4
Monoclonal
DyLight 650
CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
This antibody was developed using a recombinant protein derived from P01730, with the exact immunogen sequence remaining proprietary.
0.1 mL
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
920
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0395
50mM Sodium Borate
Mouse
Protein A purified