missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (EDU-2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11421DL594
This item is not returnable.
View return policy
Description
CD4 Monoclonal antibody specifically detects CD4 in Human, Monkey, Chimpanzee, Orangutan samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications
| CD4 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Monkey, Chimpanzee, Orangutan | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| EDU-2 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Stimulated human leukocytes | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction