missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (GK1.5), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Marke: Novus Biologicals FAB554X
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cell depletion, Costimulation of T Cells, Inhibition of T Cell Function, CyTOF
Spezifikation
| CD4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Depletion Assay, T-Cell Activation | |
| GK1.5 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cell depletion, Costimulation of T Cells, Inhibition of T Cell Function, CyTOF-ready | |
| CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Mouse CTL clone V4 | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Store at 4C in the dark. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto