missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (GK1.5), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB554MFV610
This item is not returnable.
View return policy
Description
CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cell depletion, Costimulation of T Cells, Inhibition of T Cell Function, CyTOF
Specifications
| CD4 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Depletion Assay, T-Cell Activation | |
| GK1.5 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cell depletion, Costimulation of T Cells, Inhibition of T Cell Function, CyTOF-ready | |
| CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Mouse CTL clone V4 | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction