Learn More
CD4 Antibody (RPA-T4), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-27216JF646
Product Code. 18115974
  • ONLINE EXCLUSIVE PRICE
    € 700.00
    € 661.50 / 0.10mL
    Save € 38.50
    5% Off
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CD4
PHA-stimulated human PBMC.
0.1 mL
Primary
Human
IgG1 κ
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
RPA-T4
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse
Protein G purified
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
920.0
Store at 4°C in the dark.