missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (V46P5B10*D5), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50140AF488
This item is not returnable.
View return policy
Description
CD4 Monoclonal specifically detects CD4 in Fish samples. It is validated for ELISA.
Specifications
| CD4 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD4 | |
| Ovalbumin-conjugated synthetic peptide | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Fish | |
| Purified |
| ELISA | |
| V46P5B10*D5 | |
| ELISA | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specific for three-spined stickleback CD4. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction