missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (YNB46.1.8 (Campath-9H)), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-75900DL594
This item is not returnable.
View return policy
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD4 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD4 | |
| Rat T-cell line (NB2) transfected with the human CD4 | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| Recognizes the 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| YNB46.1.8 (Campath-9H) | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction