Learn More
CD4 Monoclonal Antibody (OKT4 (OKT-4)), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 2985.00 - € 14910.00
Specifications
| Antigen | CD4 |
|---|---|
| Clone | OKT4 (OKT-4) |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay, Immunoprecipitation |
| Classification | Monoclonal |
Description
The OKT4 monoclonal antibody reacts with human CD4, a 59 kDa cell surface glycoprotein expressed by the majority of thymocytes, a subpopulation of mature T cells (T-helper cells) and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). The OKT4 antibody recognizes a different epitope than the RPA-T4 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes. Applications Reported: This OKT4 (OKT-4) antibody has been reported for use in flow cytometric analysis and immunoprecipitation. Applications Tested: This OKT4 (OKT-4) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Control, Flow Cytometry, Functional Assay, ImmunoprecipitationSpecifications
| CD4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 receptor; CD4, allele 1; cd4a; CD4mut; cell surface glycoprotein CD4; fCD4; L3T4; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); W3/25; W3/25 antigen | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
| OKT4 (OKT-4) | |
| Flow Cytometry, Functional Assay, Immunoprecipitation | |
| Functional Grade | |
| Mouse | |
| Human | |
| P01730 | |
| 920 | |
| Primary | |
| 4° C | |
| CD4 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.