missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT23 Monoclonal Antibody (OTI3F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808818S
This item is not returnable.
View return policy
Description
KRT23 Monoclonal antibody specifically detects KRT23 in Human samples. It is validated for Western BlotSpecifications
| KRT23 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZI7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7342 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3F2 | |
| Unconjugated | |
| UBP1 | |
| C HAIK1; CK23; CK-23; cytokeratin 23; cytokeratin-23; DKFZP434G032; HAIK1; histone deacetylase inducible keratin 23; hyperacetylation-inducible type I keratin; K23; Ka23; keratin 23; keratin 23 (histone deacetylase inducible); keratin 23, type I; keratin complex 1, acidic, gene 23; keratin, type I cytoskeletal 23; Keratin-23; Krt1-23; KRT23; MGC26158; type I intermediate filament cytokeratin; type I keratin KA23 | |
| Human recombinant protein fragment corresponding to amino acids 47-269 of human UBP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction