missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPNMB Monoclonal Antibody (OTI8A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807721
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a type I transmembrane glycoprotein which shows homology to the pMEL17 precursor, a melanocyte-specific protein. GPNMB shows expression in the lowly metastatic human melanoma cell lines and xenografts but does not show expression in the highly metastatic cell lines. GPNMB may be involved in growth delay and reduction of metastatic potential. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| GPNMB | |
| Monoclonal | |
| Unconjugated | |
| GPNMB | |
| HGFIN, NMB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10457 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8A10 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14956 | |
| GPNMB | |
| Full length human recombit protein of human GPNMB produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction