missing translation for 'onlineSavingsMsg'
Learn More

CD4, Mouse anti-Human, Clone: 11A1, Abnova™

Product Code. 16149339
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16149339 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16149339 Supplier Abnova Supplier No. MAB15251.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen CD4
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 11A1
Conjugate Unconjugated
Dilution Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.5% BSA, 0.02% sodium azide).
Gene CD4 molecule
Gene Alias CD4mut
Gene Symbols CD4
Host Species Mouse
Immunogen A synthetic peptide corresponding to N-terminus of human CD4.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920
Target Species Human, Mouse
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.