Learn More
CD4 Mouse anti-Rat, FITC, Clone: OX-35, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561833
Description
The OX-35 clone has been reported to react with the CD4 antigen on most thymocytes, a subpopulation of mature T lymphocytes (i.e. MHC class II-restricted T cells, including most T helper cells), monocytes, macrophages, some dendritic cells, and microglia. CD4 is an antigen coreceptor on the T-cell surface that interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through it's association with the T-cell receptor complex and protein tyrosine kinase Lck. The OX-35 clone has been reported to bind to a different epitope of CD4 than that recognized by the W3/25 and OX-38 clones.
Specifications
| CD4 | |
| Monoclonal | |
| 0.5mg/mL | |
| Cd4; CD4 antigen; p55; W3/25 antigen; T-cell surface glycoprotein CD4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Rat | |
| IgG2a κ |
| Flow Cytometry | |
| OX-35 | |
| FITC | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat T-cell blasts | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.