Learn More
CD4 Rat anti-Mouse, CoraLite 647, Clone: RM4-5, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL647-65141-100TESTS
Description
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with balb/c mouse thymocytes of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.
Specifications
| CD4 | |
| Monoclonal | |
| 5 μL/Test | |
| Rat | |
| Primary | |
| Mouse |
| Immunofluorescence, Flow Cytometry | |
| RM4-5 | |
| CoraLite 647 | |
| 100 Tests | |
| 12504 | |
| IgG2a κ |