Learn More
CD4 Rat anti-Mouse, CoraLite 647, Clone: RM4-5, Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech CL647-65141-100TESTS
Product Code. 16871146
  • € 126.00 / 100 tests
Description
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with balb/c mouse thymocytes of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.
Specifications
Specifications
CD4
Monoclonal
5 μL/Test
Rat
Primary
Mouse
Immunofluorescence, Flow Cytometry
RM4-5
CoraLite 647
100 Tests
12504
IgG2a κ