missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat anti-Mouse, Oligo, Clone: RM4-5 (also known as RM4.5), BD™ AbSeq ™
Rat Monoclonal Antibody
Brand: BD Biosciences 940108
This item is not returnable.
View return policy
Description
CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD4 | |
| Monoclonal | |
| Oligo | |
| Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3 | |
| Mouse Thymocytes (BALB/c) | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Single Cell 3' Sequencing | |
| Clone RM4-5 (also known as RM4.5) | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction