missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40 Ligand/TNFSF5 Antibody (208109), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1163C
This item is not returnable.
View return policy
Description
CD40 Ligand/TNFSF5 Monoclonal antibody specifically detects CD40 Ligand/TNFSF5 in Mouse samples. It is validated for Flow Cytometry, Neutralization, CyTOF
Specifications
| CD40 Ligand/TNFSF5 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Neutralization, CyTOF | |
| 208109 | |
| Flow Cytometry, Neutralization, CyTOF-ready | |
| CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD40 Ligand/TNFSF5, Glu61-Leu260, Accession # P27548 | |
| 0.5 mL | |
| Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| 959 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction