missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40, Mouse anti-Human, Clone: 1F11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000958-M13.100ug
This item is not returnable.
View return policy
Description
Sequence: EPPTACREKQYLINSQCCSLCQPGQKLVSDCTEFTETECLPCGESEFLDTWNRETHCHQHKYCDPNLGLRVQQKGTSETDTICTCEEGWHCTSEACESCVLHRSCSPGFGVKQIATGVSDTICEPCPVGFFSNVSSAFEKCHPWTSCETKDLVVQQAGTNKTDVVCGPQDRLRSpecifications
| CD40 | |
| Monoclonal | |
| Unconjugated | |
| CD40 molecule, TNF receptor superfamily member 5 | |
| Bp50/CDW40/MGC9013/TNFRSF5/p50 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1F11 | |
| In 1x PBS, pH 7.4 | |
| BC012419 | |
| CD40 | |
| CD40 (AAH12419, 21 a.a. ∼ 193 a.a) partial recombinant protein. | |
| RUO | |
| 958 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction