Learn More
CD40 Rat anti-Mouse, PE, Clone: 3/23, BD
Rat Monoclonal Antibody
€ 95.23 - € 313.51
Specifications
| Antigen | CD40 |
|---|---|
| Clone | 3/23 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15811399
|
BD Biosciences
561846 |
25 μg |
€ 95.30 € 95.23 / Each Save € 0.07 0.07% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15837398
|
BD Biosciences
553791 |
0.2 mg |
€ 315.00 € 313.51 / Each Save € 1.49 0.47% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 3/23 clone monoclonal antibody specifically binds to CD40, a 40-50 kDa glycoprotein expressed on B lymphocytes and other antigen-presenting cells. CD40 has been reported to be transiently expressed on activated CD4+ and CD8+ T cells and in some mouse strains, the 3/23 mAb has been reported to react with 5-10% of T lymphocytes in adult mouse, but not neonatal, spleen. CD40 plays a key role in B-cell growth and differentiation where interactions of CD40 with its ligand, CD154, are involved in the initiation, effector, and memory stages of cell-mediated immune responses. In addition, CD40 has been reported to be involved with the triggering of NK cells and NK-T cells. Ligation of CD40 with the 3/23 antibody has been reported to induce splenic B cells to express the costimulatory molecule CD86 (B7-2). In addition, although the 3/23 antibody by itself is a weak B-cell mitogen, it has been reported to synergize markedly with mitogenic anti-IgM, anti-IgD mAb or IL-4 to promote B-cell proliferation.
Specifications
| CD40 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Mouse CD40 Recombinant Protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 3/23 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.