Learn More
Novus Biologicals™ Cullin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-29486
Product Code. 30045123
  • € 571.00 / 100µg
Description
Cullin 1 Polyclonal antibody specifically detects Cullin 1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cullin 1
Polyclonal
Tris-citrate/phosphate buffer, pH 7 to 8
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunoprecipitation
Unconjugated
CUL-1, cullin 1, cullin-1, MGC149834, MGC149835
between 250 and 300
100 μg
Apoptosis, Cell Cycle and Replication
8454
Store at 4°C. Do not freeze.