missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40/TNFRSF5 Antibody (T8P2G4*A6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 369.00
Specifications
| Antigen | CD40/TNFRSF5 |
|---|---|
| Clone | T8P2G4*A6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695276
|
Novus Biologicals
NBP2-50284-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18684148
|
Novus Biologicals
NBP2-50284 |
0.1 mL |
€ 369.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| CD40/TNFRSF5 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 958 | |
| IgG2b κ | |
| Protein A purified |
| T8P2G4*A6 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
| Ovalbumin-conjugated synthetic peptide RDPVCHPLGA (C-terminal sequence) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title