missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40/TNFRSF5 Rat anti-Mouse, Clone: 1C10, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB440-100
This item is not returnable.
View return policy
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Agonist Activity, CyTOF-ready.Specifications
| CD40/TNFRSF5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD40 | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant mouse CD40/TNFRSF5 Extracellular domain | |
| 100 μg | |
| Primary | |
| Specifically binds mouse CD40/TNFRSF5 expressed on L cells. | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Agonist Activity, CyTOF | |
| 1C10 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation, Agonist Activity, CyTOF-ready | |
| B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 958 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?