missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD41/CD61 Monoclonal antibody specifically detects CD41/CD61 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Flow (Cell Surface), Immunocytochemistry
Specifications
Specifications
| Antigen | CD41/CD61 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Immunogen | Human platelets |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Research Discipline | Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgM κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?