missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD41 Monoclonal Antibody (HIP2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA119382
This item is not returnable.
View return policy
Description
This antibody reacts with an extracellular epitope on beta (b) subunit of CD41 glycoprotein (light chain; 23 kDa). CD41 is mainly expressed on platelets and megakaryocytes.
CD41, also known as platelet glycoprotein IIb or ITGA2B, is a protein composed of two subunits: a 120 kDa alpha subunit and a 23 kDa beta subunit. It interacts with CD61 (gpIIIa, integrin beta III) in the presence of calcium to form a functional adhesive protein receptor. Initially thought to be expressed exclusively on platelets and megakaryocytes, CD41 is also found on hematopoietic progenitors in the embryo, fetus, and adult, indicating its role in early stages of hematopoietic differentiation. CD41 plays a crucial role in platelet function and blood coagulation by mediating platelet aggregation. Upon blood vessel damage, CD41 binds to various adhesion molecules, including von Willebrand factor, fibrinogen, fibronectin, and vitronectin, facilitating platelet adhesion and aggregation. This receptor is essential for platelet function, contributing to the binding of several adhesion molecules and playing a significant role in hemostasis. Diseases associated with CD41 dysfunction include Glanzmann Thrombasthenia and Platelet type-16 bleeding disorder, highlighting its importance in normal platelet function and blood coagulation processes.
Specifications
| CD41 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P08514 | |
| Itga2b | |
| Human CD41b. | |
| 100 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG3 |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| HIP2 | |
| Unconjugated | |
| Itga2b | |
| AI172977; alpha IIb; alphaIIb; alphaIIb protein; BDPLT16; BDPLT2; CD41; CD41B; form 1; form 2; GP IIb; GP2B; GPalpha IIb; GPIIb; GT; GTA; HPA3; integrin alpha 2b; integrin alpha 2b (Cd41b); integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin, alpha 2B; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); ITGA2B; ITGAB; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/II Ia complex; platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3674 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction