Learn More
CD41 Rat anti-Mouse, FITC, Clone: MWREG30, BD
Rat Monoclonal Antibody
€ 118.77 - € 310.73
Specifications
| Antigen | CD41 |
|---|---|
| Clone | MWREG30 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15841399
|
BD Biosciences
561849 |
50 μg |
€ 120.00 € 118.77 / Each Save € 1.23 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15807428
|
BD Biosciences
553848 |
0.5 mg |
€ 312.00 € 310.73 / Each Save € 1.27 0.41% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MWReg30 antibody reacts with glycoprotein (gp) IIb, also called integrin αIIb chain (CD41), that associates with integrin β3 chain (gpIIIa or CD61) to form the gpIIb/IIIa (CD41/CD61) complex. CD41/CD61 is expressed on platelets, megakaryocytes, and early hematopoietic progenitors. The integrin complex binds to fibrinogen, fibronectin, vitronectin, von Willebrand factor, and thrombospondin. It is important for platelet adhesion and aggregation, and it may play a role in osteolytic tumor metastasis.
Specifications
| CD41 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Mouse Platelets | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| MWREG30 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| Integrin α IIb chain |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.