Learn More
CD42b/GPIb alpha Antibody (AK2), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63790AF405
This item is not returnable.
View return policy
Description
CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human, Mouse samples. It is validated for Flow Cytometry, ELISA.
Specifications
| CD42b/GPIb alpha | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GP1BA | |
| Made to Human CD42b | |
| 0.1 mL | |
| Immunology | |
| 2811 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, ELISA | |
| AK2 | |
| Flow Cytometry, ELISA | |
| Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognises the human CD42b cell surface antigen, also known as platelet glycoprotein GP1B. CD42b is expressed by platelets and megakaryocytes. Clone AK2 has been reported to block the binding of von Willebrand Factor (VWF) to platelets (1). | |
| Store at 4C in the dark. | |
| IgG1 |