Learn More
CD42b/GPIb alpha Antibody (AK2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-63790APC
Product Code. 18065114
  • € 620.00 / 0.10mL
Description
CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human, Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD42b/GPIb alpha
Monoclonal
APC
PBS with 0.05% Sodium Azide
GP1BA
Made to Human CD42b
0.1 mL
Immunology
2811
Human, Mouse
Purified
Flow Cytometry
AK2
Flow Cytometry
Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit
Mouse
Protein G purified
RUO
Primary
Recognises the human CD42b cell surface antigen, also known as platelet glycoprotein GP1B. CD42b is expressed by platelets and megakaryocytes. Clone AK2 has been reported to block the binding of von Willebrand Factor (VWF) to platelets (1).
Store at 4C in the dark.