missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD43 Monoclonal Antibody (W3/13)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516976
This item is not returnable.
View return policy
Description
Heat-mediated antigen retrieval is recommended for the staining of paraffin sections. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD43, also known as leukosialin or sialophorin, is a transmembrane mucin-like protein characterized by a high negative charge. It is expressed on the surface of most hematopoietic cells, excluding resting B lymphocytes. CD43 serves both adhesive and anti-adhesive functions, contributing to a repulsive barrier that can interfere with cellular adhesion while also promoting leukocyte aggregation in certain contexts. CD43 is involved in the activation of T cells, B cells, NK cells, and monocytes. Its expression decreases upon activation due to proteolytic processing. The counter-receptor for CD43 is CD169/SIGLEC-1, which is expressed on macrophages. CD43 interacts with actin-binding proteins ezrin and moesin, playing a regulatory role in remodeling T-cell morphology and regulating cell-cell interactions during lymphocyte traffic. It enhances LFA-1 adhesiveness while counteracting LFA-1 induction via other receptors. CD43 signaling induces the functionally active tumor suppressor p53 protein, but in cases of p53 and ARF deficiency, it can promote tumor proliferation and viability. CD43 is an important modulator of leukocyte functions, with dysfunctions associated with diseases such as Wiscott-Aldrich Syndrome and Adenoid Basal Cell Carcinoma.
Specifications
| CD43 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P13838 | |
| SPN | |
| Rat thymocyte membrane glycoproteins. | |
| 1 mg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| W3/13 | |
| Unconjugated | |
| SPN | |
| 3E8 antigen; A630014B01Rik; B-cell differentiation antigen LP-3; CD43; CD43 cytoplasmic tail; CD43-ct; Galactoglycoprotein; GALGP; gpL115; Leukocyte sialoglycoprotein; leukosialin; leukosianin; LOC101120462; LOW QUALITY PROTEIN: leukosialin; Lsn; Lsn1; Ly48; Ly-48; Lymphocyte antigen 48; sialophorin; sialophorin (gpL115, leukosialin, CD43); sialophorin gpL115; SPN; W3/13 antigen | |
| Mouse | |
| Protein G | |
| RUO | |
| 24796 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction