Learn More
CD43 Mouse anti-Human, Brilliant Violet 711, Clone: 1G10, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 743614
Description
The 1G10 monoclonal antibody specifically binds to CD43, a 95-135 kDa sialoglycoprotein that is highly expressed on most human leucocytes. CD43 is encoded by the SPN /gene and is also known as Sialophorin, Galactoglycoprotein (GALGP), Leukosialin, and Leukocyte sialoglycoprotein. The CD43 antigen is expressed on T cells, pre-B cells and activated B cells, NK cells and granulocytes, but is not present on resting peripheral blood B cells, red blood cells, platelets and non-hematopoietic cells. CD43 is enzymatically shed from leucocyte surfaces following activation by various stimuli. CD43 appears to be involved in intercellular interactions that regulate T, B, and NK cell functions. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections with TUF pretreatment.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications
| CD43 | |
| 1G10 | |
| Brilliant Violet 711 | |
| SPN, Sialophorin, Galactoglycoprotein/GALGP, GPL115, LEUK, Leukosialin, LSN | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.