missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTAG1B Antibody (OTI2B6), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70485MFV500
This item is not returnable.
View return policy
Description
CTAG1B Monoclonal antibody specifically detects CTAG1B in Human samples. It is validated for Flow Cytometry
Specifications
| CTAG1B | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 1485 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI2B6 | |
| Flow Cytometry | |
| Autoimmunogenic cancer/testis antigen NY-ESO-1, cancer antigen 3, cancer/testis antigen 1, cancer/testis antigen 1B, Cancer/testis antigen 6.1, CT6.1L antigen family member 2, CTAG1CTAG1A, ESO1CTAG, LAGE2, LAGE2A, LAGE2BLAGE-2, New York esophageal squamous cell carcinoma 1, NY-ESO-1 | |
| Full-length protein expressed in 293T cell transfected with human CTAG1B expression vector | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction