Learn More
CD44 Antibody (Hermes-1) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP2-22530
This item is not returnable.
View return policy
Description
CD44 Monoclonal specifically detects CD44 in Human, Mouse, Bovine, Goat, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready, Block/Neutralize.
Specifications
| Monoclonal | |
| CD44 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot 1:1 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:50, CyTOF-ready, Neutralization | |
| Protein G purified | |
| Purified |