missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Antibody (Hermes-1) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22530JF669
This item is not returnable.
View return policy
Description
CD44 Monoclonal antibody specifically detects CD44 in Bovine,Goat,Human,Mouse,Rabbit,Sheep samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,CyTOF,Neutralization,Immunocytochemistry/ Immunofluorescence
Specifications
| CD44 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| 80 - 95 kDa lymphocyte surface glycoprotein H-CAM (CD44). | |
| 0.2 mL | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Core ESC Like Genes, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 960 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, CyTOF, Neutralization, Immunocytochemistry/Immunofluorescence | |
| Hermes-1 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Goat, Human, Mouse, Rabbit, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction