missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Antibody (IM7), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
€ 497.00
Specifications
| Antigen | CD44 |
|---|---|
| Clone | IM7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Biotin |
Description
CD44 Monoclonal antibody specifically detects CD44 in Human, Mouse, Canine, Equine, Feline samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOFSpecifications
| CD44 | |
| Western Blot | |
| Biotin | |
| Rat | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Core ESC Like Genes, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 960 | |
| IgG2b | |
| Protein G purified |
| IM7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Dexamethasone-induced cells of the SJL mouse spontaneous myeloid leukemia M1 | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title