missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Antibody (VFF-17), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65535APCCY7
This item is not returnable.
View return policy
Description
CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD44 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CD44 | |
| Made to Human CD44v7/8 | |
| 0.1 mL | |
| Primary | |
| Specifically recognizes an epitope encoded by exons v7-v8 on the variant portion of human CD44. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| VFF-17 | |
| Flow Cytometry | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Mouse | |
| Protein A or G purified | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Core ESC Like Genes, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 960.0 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction