Learn More
CD44 Mouse anti-Human, Clone: HCAM/1097, Abnova™
Mouse monoclonal antibody raised against full length recombinant human CD44.
Brand: Abnova MAB13474.100ug
Description
The protein encoded by this gene is a cell-surface glycoprotein involved in cell-cell interactions, cell adhesion and migration. It is a receptor for hyaluronic acid (HA) and can also interact with other ligands, such as osteopontin, collagens, and matrix metalloproteinases (MMPs). This protein participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing, hematopoiesis, and tumor metastasis. Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full length nature of some of these variants has not been determined. Alternative splicing is the basis for the structural and functional diversity of this protein, and may be related to tumor metastasis. [provided by RefSeq]
Specifications
| CD44 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CDW44/CSPG8/ECMR-III/HCELL/IN/LHR/MC56/MDU2/MDU3/MGC10468/MIC4/MUTCH-I/Pgp1 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 960 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| HCAM/1097 | |
| Mouse monoclonal antibody raised against full length recombinant human CD44. | |
| In 10mM PBS. | |
| CD44 | |
| Recombinant protein corresponding to full length human CD44. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.