Learn More
CD44 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-15675-100UL
Description
The CD44 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CD44. This antibody recognizes human, mouse, rat antigen. The CD44 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| CD44 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Immunofluorescence | |
| CoraLite 488 | |
| 100 μL | |
| 960 | |
| IgG |