Learn More
CD44 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-15675-100UL
Product Code. 17581546
  • € 450.00 / 100µL
Description
The CD44 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CD44. This antibody recognizes human, mouse, rat antigen. The CD44 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
CD44
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Immunofluorescence
CoraLite 488
100 μL
960