missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD44, variant 6 Antibody (bivatuzumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28588-50ug
This item is not returnable.
View return policy
Description
CD44, variant 6 Monoclonal antibody specifically detects CD44, variant 6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD44, variant 6 | |
| Monoclonal | |
| Unconjugated | |
| P16070 | |
| Human | |
| Protein A purified | |
| RUO | |
| 960 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| bivatuzumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| CD44v6 | |
| 50 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Corrección del contenido de un producto
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction