missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD44, variant 6 Antibody (bivatuzumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28588-1mg
This item is not returnable.
View return policy
Description
CD44, variant 6 Monoclonal antibody specifically detects CD44, variant 6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD44, variant 6 | |
| Monoclonal | |
| Unconjugated | |
| P16070 | |
| Human | |
| Protein A purified | |
| RUO | |
| 960 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| bivatuzumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| CD44v6 | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction