missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD44var (v7) Monoclonal Antibody (VFF-9), Biotin, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ BMS117BT

Product Code. 15521387

  • € 342.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Description: Specifically recognizes an epitope encoded by exon v7 on the variant portion of human CD44. Standard CD44 (CD44s) lacks the entire variable region and is found mainly in cells of lymphohematopoietic origin. The multiple isoforms (designated CD44v; v = variant) contain alternatively spliced exons inserted within the extracellular domain. These isoforms are known as CD44v1, CD44v2, CD44v3, CD44v4, CD44v5, CD44v6, CD44v7, CD44v8, CD44v9, CD44v10, CD44v11. Applications Tested: Flow Cytometry. Purity: >95%.

CD44 cell surface antigen is a 100kDa glycoprotein widely expressed on human leucocytes, white matter of the brain and by some epithelial cells of the intestine and breast. Several isoforms of CD44 exist, including the predominant CD44H isoform detected in many normal tissues. CD44 is a receptor for hyaluronic acid (HA) and is involved in cell-cell interactions, cell adhesion and migration. CD44 also participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing. CD44 expression may be up-regulated upon some carcinomas, and it has been speculated that this may be related to metastatic potential.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD44var (v7)
Monoclonal
0.1 mg/mL
TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4
P16070
CD44
Affinity chromatography
RUO
960
2°C to 8°C
Liquid
Flow Cytometry
VFF-9
Biotin
CD44
AL133330.1; CD44; CD44 antigen; CD44 molecule (Indian blood group); CD44 splice variant; CD44 var; CD44 variant; CD44v7; CDw44; cell surface glycoprotein CD44; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; epican; Extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; H-CAM v7; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; Hermes antigen; homing function and Indian blood group system; HUTCHI; HUTCH-I; Hyaluronate receptor; IN; LHR; MC56; MDU2; MDU3; MGC10468; MIC4; MUTCH-I; PGP I; Pgp1; PGP-1; PGP-I; Phagocytic glycoprotein 1; Phagocytic glycoprotein I; soluble CD44
Mouse
100 μg
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.