missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD45.2 Monoclonal Antibody (104), Brilliant Violet™ 421, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 168.00 - € 385.00
Specifications
| Antigen | CD45.2 |
|---|---|
| Clone | 104 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17838432
|
Invitrogen™
404-0454-80 |
25 μg |
€ 168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17817912
|
Invitrogen™
404-0454-82 |
100 μg |
€ 385.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45.2 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45.2 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45.2 protein exists as multiple isoforms as a result of alternative splicing; these isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. These isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45.2 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells.Specifications
| CD45.2 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| B220; Cd45; CD45 antigen; CD45R; GP180; LCA; L-CA; leukocyte common antigen; loc; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase, receptor type, C; Ptprc; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein | |
| Ptprc | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 104 | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Mouse | |
| P06800 | |
| 19264 | |
| IgG2a κ | |
| Affinity chromatography | |
| Ptprc |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title