missing translation for 'onlineSavingsMsg'
Learn More

CD45.2 Mouse, FITC, Clone: 104, enQuireBio™

Mouse Monoclonal Antibody

€ 85.85 - € 263.00

Specifications

Antigen CD45.2
Clone 104
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15952028
View Documents Promotion Details
EnquireBio
QAB43-F-25UG
25 μg
€ 85.85
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15942028
View Documents Promotion Details
EnquireBio
QAB43-F-100UG
100 μg
€ 187.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15962028
View Documents Promotion Details
EnquireBio
QAB43-F-500UG
500 μg
€ 263.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD45.2
0.5mg/mL
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
19264
IgG2a κ
Protein A/G Purified
104
Flow Cytometry
FITC
RUO
NP_001255215
Ptprc
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications