missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Antibody (EM-05), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-44763AF488
This item is not returnable.
View return policy
Description
CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Flow (Cell Surface), CyTOF
Specifications
| CD45 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen | |
| This CD45 Antibody (EM-05) was prepared from Murine peripheral blood leukocytes. | |
| 0.1 mL | |
| Adaptive Immunity, Autoimmune Diseases, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Inflammation, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Flow Cytometry (Cell Surface) | |
| EM-05 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction