missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Antibody (HLe-1(2D1)) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 372.00
Specifications
| Antigen | CD45 |
|---|---|
| Clone | HLe-1(2D1) |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18435319
|
Novus Biologicals
NB100-2658 |
0.1 mg |
€ 372.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18447939
|
Novus Biologicals
NB100-2658-0.025mg |
0.025 mg |
€ 214.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, CyTOFSpecifications
| CD45 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Autoimmune Diseases, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Inflammation, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 5788 | |
| IgG1 | |
| Protein A purified |
| HLe-1(2D1) | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen | |
| This CD45 Antibody (HLe-1(2D1)) was prepared from Human peripheral blood mononuclear cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title