missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Antibody (YW62.3), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-65366FR
This item is not returnable.
View return policy
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD45 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PTPRC | |
| Mouse spleen cells. | |
| 0.125 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| YW62.3 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes the murine CD45 cell surface antigen, a 180-220kD glycoprotein expressed by all leucocytes. N.B. Some reactivity with human tissue has been observed. | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction