Learn More
CD45 Mouse, anti-Human, Atlantic Blue, Clone: HI30, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech AB-65109-100TESTS
Description
The CD45 antibody from Proteintech is a mouse monoclonal antibody generated with human peripheral blood leucocytes of human CD45. This antibody recognizes human antigen. The CD45 antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.
Specifications
| CD45 | |
| HI30 | |
| Atlantic Blue | |
| Mouse | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 5 μL/Test | |
| B220, CD45, CD45R, GP180, L CA, LCA, Leukocyte common antigen, LY5, PTPRC, RPTPC, T200 | |
| 100 Tests | |
| 5788 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.