missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse, PE, Clone: 2D1, enQuireBio™
Mouse Monoclonal Antibody
€ 98.45 - € 250.00
Specifications
Antigen | CD45 |
---|---|
Clone | 2D1 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932658
|
EnquireBio
QAB97-PE-25TESTS |
25 Tests |
€ 98.45
25 tests |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922658
|
EnquireBio
QAB97-PE-100TESTS |
100 Tests |
€ 250.00
100 tests |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD45 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
5788 | |
IgG1 κ | |
Protein A/G Purified |
2D1 | |
Flow Cytometry | |
PE | |
RUO | |
NP_002829 | |
PTPRC | |
Primary | |
Store at 2-4°C. |