Learn More
CD45 Mouse, PerCP, Clone: 2D1, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB97-PCP-100Tests
This item is not returnable.
View return policy
Specifications
| CD45 | |
| Monoclonal | |
| 5μL/Test | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| PTPRC | |
| Protein A/G Purified | |
| RUO | |
| 5788 | |
| IgG1 κ |
| Flow Cytometry | |
| 2D1 | |
| PerCP | |
| NP_002829 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store at 2-4°C. |