missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Rat, anti-Mouse, Biotin, Clone: 30-F11, STEMCELL Technologies
Rat Monoclonal Antibody
€ 109.35 - € 473.85
Spezifikation
| Antigen | CD45 |
|---|---|
| Clone | 30-F11 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
17188621
|
Stemcell Technologies
60030BT.1 |
50 μg |
€ 109.35
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17178621
|
Stemcell Technologies
60030BT |
500 μg |
€ 473.85
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
- The 30-F11 antibody reacts with an extracellular epitope found on all isoforms and both alloantigens (CD45.1 and CD45.2) of mouse CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets and plasma cells; expression of CD45 is lost during differentiation of these cell types
- CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation and oncogenic transformation
- The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains, one which is enzymatically active, that are involved in modulating the function of intracellular substrates such as the Src kinases Lck and Fyn
- Several isoforms of CD45 have been identified, the expression of which differs according to cell type and functional status
Specifica
| CD45 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Biotin | |
| Mouse | |
| Primary |
| 30-F11 | |
| Monoclonal | |
| Rat | |
| IgG2b κ |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.
Titolo del prodotto